2-Naphthyl boronic acid 97+%

Warning

Synonyms: 2-Naphthyl boric acidNaphthalene-2-boronic acid2-Naphthaleneboronic acid

AC35962-0010
200028-888EA 22.8 USD
200028-888
2-Naphthyl boronic acid 97+%
2-Naphthyl boronic acid
Formula: C₁₀H₉BO₂
MW: 171.99 g/mol
Melting Pt: 269…275 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00236051
CAS Number: 32316-92-0

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder and/or chunks
Infrared spectrum Conforms
HPLC ≥97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR