3-Hydroxyphenylboronic acid 97%

Warning

Synonyms: (3-Hydroxyphenyl)boronic acid(3-Hydroxyphenyl)metaboric acidDihydroxy(3-hydroxyphenyl)borane3-Hydroxybenzeneboronic acid

AC35941-0010 AC35941-2500 AC35941-0100
103512-664EA 45.9 USD
103512-664 103512-666 200023-336
3-Hydroxyphenylboronic acid 97%
3-Hydroxyphenylboronic acid
Formula: C₆H₇BO₃
MW: 137.93 g/mol
Melting Pt: 212 °C
Storage Temperature: Ambient
MDL Number: MFCD01074603
CAS Number: 87199-18-6

Order Now

Specification Test Results

Appearance Pink to grey to tan Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR