Furan-3-boronic acid 97%

Warning

Synonyms: 3-Furanylboronic acid3-Furanboronic acid

AC35940-5000 AC35940-1000
200037-494EA 18 USD
200037-494 200043-582
Furan-3-boronic acid 97%
Furan-3-boronic acid
Formula: C₄H₅BO₃
MW: 111.89 g/mol
Melting Pt: 126…128 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01319007
CAS Number: 55552-70-0

Order Now

Specification Test Results

Appearance White to yellow to beige Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR