3-Formylphenylboronic acid 97%
|
Warning
|
Synonyms:
3-Formylbenzeneboronic acid
, Dihydroxy(3-formylphenyl)borane
, (3-Formylphenyl)boronic acid
, 3-Boronobenzaldehyde
, (3-Formylphenyl)metaboric acid
AC35937-0050
AC35937-0250
AC35937-0010
103512-656EA
355
USD
103512-656
200029-302
200017-072
3-Formylphenylboronic acid 97%
3-Formylphenylboronic acid
Formula:
C₇H₇BO₃ MW: 149.94 g/mol Melting Pt: 100 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00161356 CAS Number: 87199-16-4 |
Specification Test Results
Appearance | Off-white to beige to light orange Powder or crystals and/or chunks |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...