2-Fluorophenylboronic acid 98%
|
Warning
|
Synonyms:
(2-Fluorophenyl)metaboric acid
, (2-Fluorophenyl)boronic acid
, 2-Fluorobenzeneboronic acid
, Dihydroxy(2-fluorophenyl)borane
AC35934-0250
AC35934-0010
AC35934-0050
103512-654EA
497.9
USD
103512-654
200005-052
200009-388
2-Fluorophenylboronic acid 98%
2-Fluorophenylboronic acid
Formula:
C₆H₆BFO₂ MW: 139.92 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00674013 CAS Number: 1993-03-9 |
Specification Test Results
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...