2,6-Dimethylphenylboronic acid 98%

Warning

Synonyms: Dihydroxy(2,6-dimethylphenyl)borane(2,6-Dimethylphenyl)metaboric acid(2,6-Dimethylphenyl)boronic acid2,6-Dimethylbenzeneboronic acid

AC35927-0010 AC35927-0250 AC35927-0050
200009-082EA 33.9 USD
200009-082 200016-236 103515-256
2,6-Dimethylphenylboronic acid 98%
2,6-Dimethylphenylboronic acid
Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
Melting Pt: 105 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01009693
CAS Number: 100379-00-8

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR