3,4-Difluorophenylboronic acid 97%

Warning

Synonyms: Dihydroxy(3,4-difluorophenyl)borane3,4-Difluorobenzeneboronic acid(3,4-Difluorophenyl)boronic acid(3,4-Difluorophenyl)metaboric acid

AC35916-0050 AC35916-0250
200009-666EA 63 USD
200009-666 103515-242
3,4-Difluorophenylboronic acid 97%
3,4-Difluorophenylboronic acid
Formula: C₆H₅BF₂O₂
MW: 157.91 g/mol
Melting Pt: 289…290 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00807405
CAS Number: 168267-41-2

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR