2,5-Difluorophenylboronic acid 97%

Warning

Synonyms: (2,5-Difluorophenyl)boronic acidDihydroxy(2,5-difluorophenyl)borane2,5-Difluorobenzeneboronic acid(2,5-Difluorophenyl)metaboric acid

AC35914-0010
200028-316EA 36.14 USD
200028-316
2,5-Difluorophenylboronic acid 97%
2,5-Difluorophenylboronic acid
Formula: C₆H₅BF₂O₂
MW: 157.91 g/mol
Melting Pt: 105…110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01863171
CAS Number: 193353-34-3

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR