2,3-Difluorophenylboronic acid
Synonyms:
Dihydroxy(2,3-difluorophenyl)borane
, 2,3-Difluorobenzeneboronic acid
, (2,3-Difluorophenyl)metaboric acid
AC35912-0010
200028-142EA
26.64
USD
200028-142
2,3-Difluorophenylboronic acid
2,3-Difluorophenylboronic acid
Formula:
C₆H₅BF₂O₂ MW: 157.91 g/mol |
MDL Number:
MFCD01863170 CAS Number: 121219-16-7 |
Specification Test Results
Appearance | White to light beige Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...