3,4-Dichlorophenylboronic acid 97%

Warning

AC35909-0250 AC35909-0050
200035-140EA 92.5 USD
200035-140 103515-238
3,4-Dichlorophenylboronic acid 97%
3,4-Dichlorophenylboronic acid
Formula: C₆H₅BCl₂O₂
MW: 190.82 g/mol
Melting Pt: 284…286 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01074646
CAS Number: 151169-75-4

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR