3-Acetylphenylboronic acid 96%
|
Warning
|
AC35883-0010
AC35883-0100
200016-920EA
123.82
USD
200016-920
200023-122
3-Acetylphenylboronic acid 96%
3-Acetylphenylboronic acid
Formula:
C₈H₉BO₃ MW: 163.97 g/mol Melting Pt: 212…214 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01074678 CAS Number: 204841-19-0 |
Specification Test Results
Appearance | White to light yellow Powder |
Infrared spectrum | Conforms |
HPLC | ≥95 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...