5-Bromo-2-hydroxypyridine 99%

Warning

Synonyms: 3-Bromo-6-pyridinol5-Bromo-2-pyridinol5-Bromopyridin-2-ol3-Bromo-6-hydroxypyridine

AC35764-0250 AC35764-0050
200005-070EA 202 USD
200005-070 200009-412
5-Bromo-2-hydroxypyridine 99%
5-Bromo-2-hydroxypyridine
Formula: C₅H₄BrNO
MW: 174 g/mol
Melting Pt: 179…182 °C
MDL Number: MFCD00234042
CAS Number: 13466-38-1

Order Now

Specification Test Results

Appearance Off-white to yellow-brown Crystalline powder
Infrared spectrum Conforms
Melting point 179 to 182 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR