Cycloheximide 95%

Danger

Synonyms: 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,Naramycin AActidione

AC35742-0250 AC35742-0050 AC35742-0010
103527-822EA 671 USD
103527-822 200063-764 200063-762
Cycloheximide 95%
Cycloheximide
Formula: C₁₅H₂₃NO₄
MW: 281.35 g/mol
Melting Pt: 107…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00082346
CAS Number: 66-81-9
EINECS: 200-636-0
UN: 2811
ADR: 6.1,I
https://us.vwr.com/stibo/web/std.lang.all/89/03/2298903.jpg

Order Now

Specification Test Results

Appearance White to light beige Powder
Infrared spectrum Conforms
Melting point 108 to 118 °C
HPLC ≥94 %
Specific optical rotation -25° to -32° (20 °C, 589 nm) (c=1, CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR