1-Hexadecyl-3-methylimidazolium chloride monohydrate 98%

Warning

Synonyms: 1-Hexadecyl-3-methyl-1H-imidazol-3-ium chloride hydrate

AC35665-0100
200004-746EA 265.38 USD
200004-746
1-Hexadecyl-3-methylimidazolium chloride monohydrate 98%
1-Hexadecyl-3-methylimidazolium chloride monohydrate
Formula: C₂₀H₃₉ClN₂·1H₂O
MW: 361.01 g/mol
MDL Number: MFCD07773044
CAS Number: 404001-62-3

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder and/or chunks
Infrared spectrum Conforms
Titration Argentometric ≥97.5 %
Water 3 to 5 % (K.F.)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR