5-Iodoanthranilic acid 98%

Warning

Synonyms: 2-Amino-5-iodobenzoic acid

AC35544-0250 AC35544-1000
200009-146EA 94.1 USD
200009-146 103513-240
5-Iodoanthranilic acid 98%
5-Iodoanthranilic acid
Formula: C₇H₆INO₂
MW: 263.03 g/mol
Melting Pt: 219…221 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007849
CAS Number: 5326-47-6
EINECS: 226-205-7

Order Now

Specification Test Results

Appearance Beige to grey to purple Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with TBAH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR