Dibutyl maleate 97%

Warning

Synonyms: Maleic acid dibutyl ester

AC35310-0010 AC35310-2500
200052-526EA 25.8 USD
200052-526 200018-734
Dibutyl maleate 97%
Dibutyl maleate
Formula: C₁₂H₂₀O₄
MW: 228.29 g/mol
Boiling Pt: 274…277 °C
Density: 0.99 g/cm³
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD00009447
CAS Number: 105-76-0
EINECS: 203-328-4
UN: 3082

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4460 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR