Tetraoctylammonium bromide 98%

Warning

Synonyms: Tetra-n-octylammonium bromide

AC35210-0250 AC35210-0050
200014-778EA 29.1 USD
200014-778 200003-500
Tetraoctylammonium bromide 98%
Tetraoctylammonium bromide
Formula: C₃₂H₆₈BrN
MW: 546.8 g/mol
Melting Pt: 95…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00011863
CAS Number: 14866-33-2
EINECS: 238-936-9

Order Now

Specification Test Results

Appearance White to cream Powder or flakes
Infrared spectrum Conforms
Melting point 95 to 99 °C
Titration Argentometric ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR