3,5-Bis(trifluoromethyl)phenylboronic acid 95%

Warning

Synonyms: 3,5-Bis(trifluoromethyl)benzeneboronic acid(3,5-Bis(trifluoromethyl)phenyl)boronic acidDihydroxy(3,5-bis(trifluoromethyl)phenyl)borane(3,5-Bis(trifluoromethyl)phenyl)metaboric acid

AC35136-0010 AC35136-0050
200009-688EA 32.3 USD
200009-688 200009-690
3,5-Bis(trifluoromethyl)phenylboronic acid 95%
3,5-Bis(trifluoromethyl)phenylboronic acid
Formula: C₈H₅BF₆O₂
MW: 257.93 g/mol
Melting Pt: 219…226 °C
MDL Number: MFCD00051850
CAS Number: 73852-19-4

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Melting point 219 to 226 °C
Titration with NaOH 94 to 108 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR