2,5-Dimethylbenzylamine 98%

Danger

Synonyms: (2,5-Dimethylphenyl)methanamine

AC35086-0010 AC35086-0050
200022-262EA 38 USD
200022-262 200009-038
2,5-Dimethylbenzylamine 98%
2,5-Dimethylbenzylamine
Formula: C₉H₁₃N
MW: 135.21 g/mol
MDL Number: MFCD00039780
CAS Number: 93-48-1
EINECS: 202-250-8
UN: 2735

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.5350 to 1.5380 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR