2,6-Dichlorobenzylamine 98%

Danger

Synonyms: (2,6-Dichlorophenyl)methanamine

AC35074-0050 AC35074-0250
200009-056EA 60.3 USD
200009-056 200009-058
2,6-Dichlorobenzylamine 98%
2,6-Dichlorobenzylamine
Formula: C₇H₇Cl₂N
MW: 176.04 g/mol
MDL Number: MFCD00047928
CAS Number: 6575-27-5
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5770 to 1.5790 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR