Celite® 545, filter aid
Synonyms:
Diatomaceous earth, flux-calcined
AC34967-0025
AC34967-5000
200003-466EA
94.9
USD
200003-466
200003-468
Celite® 545, filter aid
Celite® 545, filter aid
Formula:
SiO₂ MW: 60.08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00132803 CAS Number: 68855-54-9 EINECS: 272-489-0 |
Specification Test Results
Appearance | White to almost white Powder |
Particle size | ≤25 % (Retained on 150 mesh) |
Wet density | 255 to 370 g/l |
pH | 9.0 to 11.0 (10 % slurry) |
Permeability | 2.6 to 6.5 Darcy |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...