4-Pyridone 95%

Danger

Synonyms: 1,4-Dihydro-4-PyridinonePyridin-4-olγ-Pyridone4-Hydroxypyridine4-Pyridinol

AC34883-5000 AC34883-1000 AC34883-0250
200010-478EA 43 USD
200010-478 200010-476 103513-938
4-Pyridone 95%
4-Pyridone
Formula: C₅H₅NO
MW: 95.1 g/mol
Boiling Pt: 230…235 °C (12 mmHg)
Melting Pt: 140…147 °C
Flash Pt: 221 °C
Storage Temperature: Ambient
MDL Number: MFCD00006419
CAS Number: 626-64-2
EINECS: 210-958-3

Order Now

Specification Test Results

Appearance White to yellow to beige to light brown Powder and/or chunks
Infrared spectrum Conforms
Melting point 139 to 151 °C
HPLC ≥94 %
Titration with HClO4 ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR