Toluidine blue O, pure

Synonyms: Tolonium chloride 3-Amino-7-(dimethylamino)-2-methyl-5-phenothiazinium chloride

AC34860-1000 AC34860-0050
102777-068EA 317 USD
102777-068 200007-908
Toluidine blue O, pure
Toluidine blue O
Formula: C₁₅H₁₆ClN₃S
MW: 305.83 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011934
CAS Number: 92-31-9
EINECS: 202-146-2
Merck Index: 12,09658

Order Now

Specification Test Results

Appearance Dark green Powder
Infrared spectrum Conforms
UV spectrum Complies

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR