Potassium carbonate sesquihydrate 98.5% ACS

Warning

AC34720-0025
103518-728EA 255 USD
103518-728
Potassium carbonate sesquihydrate 98.5% ACS
Potassium carbonate sesquihydrate
Formula: CK₂O₃·1.5H₂O
MW: 165.23 g/mol
Melting Pt: 891 °C
Storage Temperature: Ambient
MDL Number: MFCD00149925
CAS Number: 6381-79-9
EINECS: 209-529-3

Order Now

Specification Test Results

Appearance White Crystalline powder
Titration with HCl 98.5 to 101.0 %
Chloride (Cl) ≤0.003 %
Phosphate (PO4) ≤0.001 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤0.002 %
Sodium (Na) ≤0.02 %
Heavy metals (ICP-OES) ≤5 ppm
Silicon dioxide (SiO2) ≤0.005 %
Sulfur compounds (as SO4) ≤0.004 %
Insoluble matter ≤0.01 %
Loss on heating 14.0 % to 16.5 % (285 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR