4-Bromobenzyl chloride 98%

Danger

Synonyms: α-Chloro-4-bromotoluene1-(Chloromethyl)-4-bromobenzene4-Bromo-α-chlorotoluene1-Bromo-4-(chloromethyl)benzene

AC34675-0050 AC34675-0250
200010-266EA 51.9 USD
200010-266 200041-838
4-Bromobenzyl chloride 98%
4-Bromobenzyl chloride
Formula: C₇H₆BrCl
MW: 205.48 g/mol
Boiling Pt: 136…139 °C (27 mmHg)
Melting Pt: 36…40 °C
MDL Number: MFCD00040867
CAS Number: 589-17-3
EINECS: 209-638-6
UN: 3261
ADR: 8,II
Merck Index: 13,01394

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Melting point 36 to 40 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR