3,4-Dimethylbenzaldehyde 97%

AC34606-0050 AC34606-0250
200041-378EA 273 USD
200041-378 200035-160
3,4-Dimethylbenzaldehyde 97%
3,4-Dimethylbenzaldehyde
Formula: C₉H₁₀O
MW: 134.18 g/mol
Boiling Pt: 68 °C (0.8 mmHg)
Density: 1.012 g/cm³
MDL Number: MFCD00016612
CAS Number: 5973-71-7
EINECS: 227-770-2

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5490 to 1.5510 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR