2',6'-Dichloro-3'-fluoroacetophenone 97%

Warning

AC34551-0250
200034-466EA 127.7 USD
200034-466
2',6'-Dichloro-3'-fluoroacetophenone 97%
2',6'-Dichloro-3'-fluoroacetophenone
Formula: C₈H₅Cl₂FO
MW: 207.03 g/mol
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD02093760
CAS Number: 290835-85-7

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5275 to 1.5295 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR