Methylcyclopentane 95%

Danger

AC34524-0250 AC34524-5000 AC34524-0025
200013-368EA 144 USD
200013-368 200043-968 103527-738
Methylcyclopentane 95%
Methylcyclopentane
Formula: C₆H₁₂
MW: 84.16 g/mol
Boiling Pt: 71 °C
Melting Pt: –142.4 °C
Density: 0.748 g/cm³
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00001382
CAS Number: 96-37-7
EINECS: 202-503-2
UN: 2298
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4070 to 1.4110 (20 °C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR