2-Methylpyridin-3-ol 99%

Danger

Synonyms: 3-Hydroxy-2-methylpyridine

AC34522-0050
200017-084EA 86.2 USD
200017-084
2-Methylpyridin-3-ol 99%
2-Methylpyridin-3-ol
Formula: C₆H₇NO
MW: 109.13 g/mol
Melting Pt: 167…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00082538
CAS Number: 1121-25-1
EINECS: 214-327-3

Order Now

Specification Test Results

Appearance Beige to light brown Powder
Infrared spectrum Conforms
Melting point 167 to 171 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR