4-Chlorophenylboronic acid 97%

Warning

Synonyms: (4-Chlorophenyl)metaboric acidDihydroxy(4-chlorophenyl)borane(4-Chlorophenyl)boronic acid4-Chlorobenzeneboronic acid

AC34441-0050 AC34441-0250 AC34441-0010
200010-356EA 59.6 USD
200010-356 103513-882 200029-754
4-Chlorophenylboronic acid 97%
4-Chlorophenylboronic acid
Formula: C₆H₆BClO₂
MW: 156.38 g/mol
Melting Pt: 259…262 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00039137
CAS Number: 1679-18-1
EINECS: 216-845-5

Order Now

Specification Test Results

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR