2-Chloro-5-methylpyridine 94%

Warning

Synonyms: 2-Chloro-5-picoline

AC34403-0250 AC34403-0050
200028-630EA 40.45 USD
200028-630 200028-628
2-Chloro-5-methylpyridine 94%
2-Chloro-5-methylpyridine
Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 97 °C (30 mmHg)
Density: 1.169 g/cm³
Flash Pt: 90 °C
MDL Number: MFCD00792460
CAS Number: 18368-64-4
EINECS: 418-050-0

Order Now

Specification Test Results

Appearance Clear light yellow to straw yellow to pink-orange Liquid
Infrared spectrum Conforms
Refractive index 1.5260 to 1.5300 (20 °C, 589 nm)
GC ≥92 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR