1-Butanesulfonic acid sodium salt 99+% for HPLC

Warning

Synonyms: Butane-1-sulphonic acid sodium saltButane-1-sulfonic acid sodium saltSodium 1-butanesulphonateSodium 1-butansulfonateSodium 1-butansulphonateSodium 1-butanesulfonateSodium butane-1-sulfonateSodium butane-1-sulphonate

AC34380-0250
103518-724EA 264 USD
103518-724
1-Butanesulfonic acid sodium salt 99+% for HPLC
1-Butansulphonic acid sodium salt
Formula: C₄H₉NaO₃S
MW: 160.17 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007540
CAS Number: 2386-54-1
EINECS: 219-201-1

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤1 % (1 g, 105 °C, 2 hrs)
Solubility (10 % in water) Clear to slightly hazy colorless
UV (5.5 % in water, pH 3) 1 cm cell
UV at 210 nm A: ≤1
UV at 220 nm A: ≤0.1
UV at 250 nm A: ≤0.04
UV at 280 nm A: ≤0.04
Titration after Ion exchange ≥99.0 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR