2,3-Dimethylthiophene 97%

Danger

AC34227-0050 AC34227-0010
200028-164EA 126 USD
200028-164 200016-052
2,3-Dimethylthiophene 97%
2,3-Dimethylthiophene
Formula: C₆H₈S
MW: 112.2 g/mol
Boiling Pt: 141.6 °C (1013 hPa)
Melting Pt: –49 °C
Flash Pt: >1 °C
Storage Temperature: Ambient
MDL Number: MFCD00130081
CAS Number: 632-16-6
EINECS: 211-170-2
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5170 to 1.5210 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR