3-Aminobenzamide 98%

Warning

Synonyms: m-AminobenzamideINO-1001

AC33908-0010 AC33908-0050 AC33908-0250
200016-938EA 85.7 USD
200016-938 200029-186 103465-258
3-Aminobenzamide 98%
3-Aminobenzamide
Formula: C₇H₈N₂O
MW: 136.15 g/mol
Melting Pt: 111…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00007989
CAS Number: 3544-24-9
EINECS: 222-586-9

Order Now

Specification Test Results

Appearance Beige Powder
Infrared spectrum Conforms
Melting point 111 to 115 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR