2-Methyl-1-butene-3-yne 98%

Danger

Synonyms: 2-Methylbut-1-en-3-yne

AC33648-0100 AC33648-0500
200063-714EA 73 USD
200063-714 103527-712
2-Methyl-1-butene-3-yne 98%
2-Methyl-1-butene-3-yne
Formula: C₅H₆
MW: 66.1 g/mol
Boiling Pt: 32 °C
Melting Pt: –113 °C
Density: 0.69 g/cm³
Flash Pt: –45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008599
CAS Number: 78-80-8
EINECS: 201-144-9
UN: 3295
ADR: 3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR