1,2-Epoxyhexane 97%
|
Danger
|
Synonyms:
Butyloxirane
AC33555-0250
AC33555-1000
AC33555-0050
200015-544EA
56.5
USD
200015-544
103527-710
200027-592
1,2-Epoxyhexane 97%
1,2-Epoxyhexane
Formula:
C₆H₁₂O Boiling Pt: 118…120 °C (760 mmHg) Density: 0.83 g/cm³ Flash Pt: 15 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005154 CAS Number: 1436-34-6 EINECS: 215-864-6 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...