5-Methoxy-2-nitrophenol 98%

Danger

AC33544-0010 AC33544-0050
200003-402EA 131 USD
200003-402 103529-586
5-Methoxy-2-nitrophenol 98%
5-Methoxy-2-nitrophenol
Formula: C₇H₇NO₄
Melting Pt: 90…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00100932
CAS Number: 704-14-3
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR