4,5-Dimethoxyanthranilic acid 98%

Warning

Synonyms: 2-Amino-4,5-dimethoxybenzoic acid6-Aminoveratric acid

AC33501-0050 AC33501-1000 AC33501-0250
200009-128EA 74.4 USD
200009-128 200046-644 103513-690
4,5-Dimethoxyanthranilic acid 98%
4,5-Dimethoxyanthranilic acid
Formula: C₉H₁₁NO₄
MW: 197.19 g/mol
Melting Pt: 156…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00011671
CAS Number: 5653-40-7
EINECS: 227-095-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR