1-Naphthyl butyrate 98%

AC33495-0010
200015-864EA 62.2 USD
200015-864
1-Naphthyl butyrate 98%
1-Naphthyl butyrate
Formula: C₁₄H₁₄O₂
MW: 214.26 g/mol
Boiling Pt: 125…130 °C (1 mmHg)
Density: 1.09 g/cm³
Flash Pt: 62 °C
Storage Temperature: Ambient
MDL Number: MFCD00003923
CAS Number: 3121-70-8
EINECS: 221-500-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR