1,2-Benzenedithiol 96%

Warning

Synonyms: Benzene-1,2-dithiol

AC33492-0050 AC33492-0010
103513-682EA 466 USD
103513-682 200015-518
1,2-Benzenedithiol 96%
1,2-Benzenedithiol
Formula: C₆H₆S₂
MW: 142.25 g/mol
Melting Pt: 27…28 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004835
CAS Number: 17534-15-5
EINECS: 241-530-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR