Sodium octanoate 98%

Warning

Synonyms: Caprylic acid sodium acidOctanoic acid sodium acidSodium n-OctanoateSodium caprylate

AC33460-1000 AC33460-0025 AC33460-5000
200026-780EA 216 USD
200026-780 200014-468 103513-668
Sodium octanoate 98%
Sodium octanoate
Formula: C₈H₁₅NaO₂
MW: 166.2 g/mol
Melting Pt: >225 °C
Storage Temperature: Ambient
MDL Number: MFCD00058511
CAS Number: 1984-06-1
EINECS: 217-850-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR