6-Methoxy-1-indanone 98%

Warning

AC33407-0250 AC33407-0010 AC33407-0050
200048-308EA 40.9 USD
200048-308 200010-806 103532-502
6-Methoxy-1-indanone 98%
6-Methoxy-1-indanone
Formula: C₁₀H₁₀O₂
MW: 162.19 g/mol
Boiling Pt: 270 °C (1013 hPa)
Melting Pt: 107…111 °C
Storage Temperature: Ambient
MDL Number: MFCD00021232
CAS Number: 13623-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR