(±)-2-Decanol 98%

Synonyms: (±)-2-Hydroxy decane(±)-Octyl methyl carbinol2-Decanol(±)-Methyl octyl carbinol

AC33160-0250 AC33160-0050
200046-812EA 25.8 USD
200046-812 200022-610
(±)-2-Decanol 98%
(±)-2-Decanol
Formula: C₁₀H₂₂O
MW: 158.28 g/mol
Boiling Pt: 211 °C (1013 hPa)
Melting Pt: –5 °C
Density: 0.82 g/cm³
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00004594
CAS Number: 1120-06-5
EINECS: 214-296-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR