4-Methyl-2-pentanone 99+%, Electronic Grade residue-free

Danger

Synonyms: Methyl Isobutyl KetoneIsobutyl Methyl KetoneMethyl iso-Butyl Ketone

AC32792-0025 AC32792-0010
200010-540EA 251.54 USD
200010-540 200005-494
4-Methyl-2-pentanone 99+%, Electronic Grade residue-free
4-Methyl-2-pentanone
Formula: (CH₃)₂CHCH₂COCH₃
MW: 100.16 g/mol
Boiling Pt: 117.4 °C (1013 hPa)
Melting Pt: –84 °C
Density: 0.8 g/cm³
Flash Pt: 14 °C
Storage Temperature: Ambient
MDL Number: MFCD00008938
CAS Number: 108-10-1
EINECS: 203-550-1
UN: 1245
ADR: 3,II
Merck Index: 13,05227
https://us.vwr.com/stibo/web/std.lang.all/93/40/739340.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR