n-Hexane 97% for electronic use (MOS) residue-free

Danger

Synonyms: HexanenHexaneHexyl hydride

AC32789-0010
200007-140EA 80.7 USD
200007-140
n-Hexane 97% for electronic use (MOS) residue-free
n-Hexane
Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.659 g/cm³
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3,II
Merck Index: 13,04712
https://us.vwr.com/stibo/web/std.lang.all/70/18/737018.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR