Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) 99+% for biochemistry

Warning

Synonyms: 2-[Bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diolBis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane (BIS-TRIS)2-(Bis(2-hydroxyethyl)amino)-2-(hydroxymethyl)propane-1,3-diol2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol

AC32772-1000 AC32772-5000
103527-662EA 373 USD
103527-662 200003-346
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) 99+% for biochemistry
Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane (BIS-TRIS)
Formula: C₈H₁₉NO₅
MW: 209.24 g/mol
Melting Pt: 103…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00002853
CAS Number: 6976-37-0
EINECS: 230-237-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR