HEPES sodium salt 99+% for molecular biology DNase-, RNase-, protease-free

Synonyms: N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulphonic acid) sodium saltSodium 2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulphonateSodium 4-(2-hydroxyethyl)piperazin-1-ylethanesulphonateSodium 2-(4-(2-hydroxyethyl)piperazin-1-yl)ethanesulfonateSodium 4-(2-hydroxyethyl)piperazin-1-ylethanesulfonate4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt4-(2-Hydroxyethyl)piperazine-1-ethanesulphonic acid sodium saltN-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid) sodium salt

AC32726-1000 AC32726-5000
200006-614EA 355 USD
200006-614 103509-996
HEPES sodium salt 99+% for molecular biology DNase-, RNase-, protease-free
HEPES sodium salt
Formula: C₈H₁₇N₂NaO₄S
MW: 260.29 g/mol
Melting Pt: 248 °C
Storage Temperature: Ambient
MDL Number: MFCD00036463
CAS Number: 75277-39-3
EINECS: 278-169-7
https://us.vwr.com/stibo/web/std.lang.all/61/70/18366170.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR