EDTA tetrasodium salt tetrahydrate 99.5% for molecular biology DNase-, RNase-, protease-free

Danger

AC32721-1000
200057-568EA 52.9 USD
200057-568
EDTA tetrasodium salt tetrahydrate 99.5% for molecular biology DNase-, RNase-, protease-free
EDTA tetrasodium salt tetrahydrate
Formula: C₁₀H₁₂N₂O₈Na₄·4H₂O
MW: 452.23 g/mol
Melting Pt: > 300 °C
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00150027
CAS Number: 13235-36-4
EINECS: 200-573-9
Merck Index: 12,03557

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR