4-Iodo-3-nitrotoluene ≥97%

Warning

Synonyms: 1-Iodo-4-methyl-2-nitrobenzene

AC32630-0050
200047-940EA 95 USD
200047-940
4-Iodo-3-nitrotoluene ≥97%
4-Iodo-3-nitrotoluene
Formula: C₇H₆INO₂
MW: 263.03 g/mol
Melting Pt: 55 °C
MDL Number: MFCD00024196
CAS Number: 5326-39-6

Order Now

Specification Test Results

Appearance Orange-brown Glistening powder
Infrared spectrum Conforms
Melting point 50°C to 55°C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR