3-Bromopropionic acid 97%

Danger

Synonyms: 3-Bromopropanoic acid

AC32577-0100 AC32577-1000 AC32577-5000
200009-836EA 45.9 USD
200009-836 200009-838 103532-458
3-Bromopropionic acid 97%
3-Bromopropionic acid
Formula: BrH₂CCH₂COOH
MW: 152.98 g/mol
Boiling Pt: 140…142 °C (45 mmHg)
Melting Pt: 57…63 °C
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00002763
CAS Number: 590-92-1
EINECS: 209-694-1
UN: 3261
ADR: 8,II
Merck Index: 13,01418

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR